The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004179.2 | NP_004170.1 | 2 - 3 | 327 | 104 |
GenBank mRNA | BC106739.1 | - | 2 - 3 | 301 | 104 |
BP423529.1 | - | 3 - 4 | 396 | 104 | |
X52836.1 | - | 2 - 3 | 301 | 104 |