The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031946.6 | NP_114152.3 | 12 - 13 | 1992 | 136 |
XM_005249943.2 | XP_005250000.1 | 12 - 13 | 1567 | 136 | |
XM_011515780.1 | XP_011514082.1 | 12 - 13 | 1604 | 136 | |
XM_017011732.1 | XP_016867221.1 | 10 - 11 | 1182 | 136 | |
GenBank mRNA | AF359283.3 | - | 13 - 14 | 1567 | 136 |
AF413079.1 | - | 12 - 13 | 1567 | 136 | |
AK096352.1 | - | 7 - 8 | 961 | 136 | |
BC037543.1 | - | 2 - 3 | 198 | 136 |