Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZNF252P-AS1
- Entrez Gene ID
-
286103
- Gene Name
- ZNF252P antisense RNA 1
- Gene Aliases
-
C8orf77
- Chromosome Location
-
Chr.8: 145002811 - 145006046 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026974.1 |
|
1 - 1 |
657 |
141 |
GenBank mRNA |
AK092777.1 |
- |
1 - 1 |
482 |
141 |
BC046439.1 |
- |
1 - 1 |
657 |
141 |
BC122544.1 |
- |
1 - 1 |
631 |
141 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA919539.1 |
- |
1 |
483 |
|
Chr. 8 - 145002986 - 145003562 [+] on Build GRCh38 |
141 |
GenBankmRNA |
DA497260.1 |
- |
1 |
455 |
|
Chr. 8 - 145003013 - 145003579 [+] on Build GRCh38 |
141 |
GenBankmRNA |
DA752317.1 |
- |
1 |
451 |
|
Chr. 8 - 145003017 - 145003587 [+] on Build GRCh38 |
141 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger protein 252, pseudogene |
ZNF252P |
ZNF252 |
-87.0
|
Anti-sense |
Chr. - 144973589 - 145002899 on Build GRCh38 |
Downstream |
chromosome 8 open reading frame 33 |
C8orf33 |
|
48.9kb
|
Sense |
Chr. - 145052438 - 145056030 on Build GRCh38 |
View Full Product Details