The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114091.2 | NP_001107563.1 | 14 - 15 | 2065 | 158 |
NM_001256.4 | NP_001247.3 | 14 - 15 | 2047 | 158 | |
NM_001293089.1 | NP_001280018.1 | 14 - 15 | 2044 | 158 | |
NM_001293091.1 | NP_001280020.1 | 13 - 14 | 1899 | 158 | |
XM_011525546.2 | XP_011523848.1 | 14 - 15 | 2049 | 158 | |
XM_011525547.2 | XP_011523849.1 | 14 - 15 | 2052 | 158 | |
XM_011525548.2 | XP_011523850.1 | 13 - 14 | 1937 | 158 | |
XM_011525549.2 | XP_011523851.1 | 13 - 14 | 2060 | 158 | |
XM_011525550.2 | XP_011523852.1 | 12 - 13 | 1747 | 158 | |
XM_017025481.1 | XP_016880970.1 | 14 - 15 | 2049 | 158 | |
XM_017025482.1 | XP_016880971.1 | 14 - 15 | 2034 | 158 | |
XM_017025483.1 | XP_016880972.1 | 14 - 15 | 2031 | 158 | |
XM_017025484.1 | XP_016880973.1 | 13 - 14 | 1912 | 158 | |
XM_017025485.1 | XP_016880974.1 | 12 - 13 | 1790 | 158 | |
XM_017025486.1 | XP_016880975.1 | 12 - 13 | 1707 | 158 | |
XM_017025487.1 | XP_016880976.1 | 13 - 14 | 1918 | 158 | |
XM_017025488.1 | XP_016880977.1 | 14 - 15 | 2014 | 158 | |
GenBank mRNA | AK296881.1 | - | 13 - 14 | 1839 | 158 |
BC011656.2 | - | 14 - 15 | 1929 | 158 | |
S78234.1 | - | 14 - 15 | 2007 | 158 | |
U00001.1 | - | 14 - 15 | 1962 | 158 |