The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025235.1 | NP_001020406.1 | 1 - 2 | 69 | |
XM_011520338.1 | XP_011518640.1 | 1 - 2 | 69 | ||
GenBank mRNA | AL543667.3 | - | 1 - 2 | 134 | 70 |
BM555783.1 | - | - | 69 |