The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024940.7 | NP_079216.4 | 19 - 20 | 2185 | 151 |
XM_011544661.1 | XP_011542963.1 | 19 - 20 | 2122 | 151 | |
XM_011544662.1 | XP_011542964.1 | 19 - 20 | 2122 | 151 | |
XM_011544663.2 | XP_011542965.1 | 19 - 20 | 2122 | 151 | |
XM_017013865.1 | XP_016869354.1 | 19 - 20 | 1974 | 151 | |
XM_017013866.1 | XP_016869355.1 | 17 - 18 | 3079 | 151 | |
XM_017013867.1 | XP_016869356.1 | 12 - 13 | 1512 | 151 | |
XM_017013868.1 | XP_016869357.1 | 13 - 14 | 1456 | 151 | |
GenBank mRNA | AK123607.1 | - | 19 - 20 | 2118 | 151 |
BC137175.1 | - | 19 - 20 | 2043 | 151 | |
BC137176.1 | - | 19 - 20 | 2043 | 151 | |
BX648352.1 | - | 2 - 3 | 218 | 151 | |
CR749353.1 | - | 11 - 12 | 1326 | 151 | |
CR933718.1 | - | 7 - 8 | 759 | 151 |