Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOCK5
Entrez Gene ID
80005
Gene Name
dedicator of cytokinesis 5
Gene Aliases
-
Chromosome Location
Chr.8: 25184723 - 25415716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024940.7 NP_079216.4 3 - 4 358 92
XM_011544661.1 XP_011542963.1 3 - 4 295 92
XM_011544662.1 XP_011542964.1 3 - 4 295 92
XM_011544663.2 XP_011542965.1 3 - 4 295 92
XM_017013865.1 XP_016869354.1 3 - 4 147 92
GenBank mRNA AK123607.1 - 3 - 4 295 92
BC011877.2 - 3 - 4 309 92
BC137175.1 - 3 - 4 216 92
BC137176.1 - 3 - 4 216 92

View Full Product Details