Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCDC180
- Entrez Gene ID
-
100499483
- Gene Name
- coiled-coil domain containing 180
- Gene Aliases
-
BDAG1, C9orf174, KIAA1529
- Chromosome Location
-
Chr.9: 97307628 - 97377295 on Build GRCh38
- Gene Symbol
- LOC100499484-C9ORF174
- Entrez Gene ID
-
57653
- Gene Name
- LOC100499484-C9orf174 readthrough
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 97238426 - 97377295 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_036527.1 |
|
28 - 29 |
3098 |
70 |
NR_036528.1 |
|
28 - 29 |
3098 |
70 |
NR_036529.1 |
|
25 - 26 |
2658 |
70 |
View Full Product Details