The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308401.1 | NP_001295330.1 | 3 - 4 | 1254 | 98 |
NM_022840.4 | NP_073751.3 | 3 - 4 | 1254 | 98 | |
XM_005258132.3 | XP_005258189.1 | 3 - 4 | 1251 | 98 | |
XM_005258133.2 | XP_005258190.1 | 3 - 4 | 293 | 98 | |
XM_011525730.2 | XP_011524032.1 | 3 - 4 | 1250 | 98 | |
GenBank mRNA | AK290001.1 | - | 3 - 4 | 1175 | 98 |
BC111020.1 | - | 3 - 4 | 1237 | 98 | |
BC136766.1 | - | 3 - 4 | 1000 | 98 | |
BC136767.1 | - | 3 - 4 | 1001 | 98 | |
BX647373.1 | - | 2 - 3 | 858 | 98 |