The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308401.1 | NP_001295330.1 | 4 - 5 | 1620 | 109 |
NM_022840.4 | NP_073751.3 | 4 - 5 | 1620 | 109 | |
XM_005258132.3 | XP_005258189.1 | 4 - 5 | 1617 | 109 | |
XM_005258133.2 | XP_005258190.1 | 4 - 5 | 659 | 109 | |
XM_011525730.2 | XP_011524032.1 | 4 - 5 | 1616 | 109 | |
GenBank mRNA | AK026670.1 | - | 2 - 3 | 420 | 109 |
AK290001.1 | - | 4 - 5 | 1541 | 109 | |
AL834296.1 | - | 1 - 2 | 146 | 109 | |
BC111020.1 | - | 4 - 5 | 1603 | 109 | |
BC136766.1 | - | 4 - 5 | 1366 | 109 | |
BC136767.1 | - | 4 - 5 | 1367 | 109 |