The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308401.1 | NP_001295330.1 | 5 - 6 | 1686 | 124 |
NM_022840.4 | NP_073751.3 | 5 - 6 | 1686 | 124 | |
XM_005258132.3 | XP_005258189.1 | 5 - 6 | 1683 | 124 | |
XM_005258133.2 | XP_005258190.1 | 5 - 6 | 725 | 124 | |
GenBank mRNA | AK022584.1 | - | 2 - 3 | 88 | 124 |
AK026670.1 | - | 3 - 4 | 486 | 124 | |
AL834296.1 | - | 2 - 3 | 212 | 124 | |
BC111020.1 | - | 5 - 6 | 1669 | 124 | |
BC136766.1 | - | 5 - 6 | 1432 | 124 | |
BC136767.1 | - | 5 - 6 | 1433 | 124 | |
BX647373.1 | - | 3 - 4 | 3193 | 124 |