Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD84
Entrez Gene ID
8832
Gene Name
CD84 molecule
Gene Aliases
LY9B, SLAMF5, hCD84, mCD84
Chromosome Location
Chr.1: 160541094 - 160579516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184881.1 NP_001171810.1 4 - 5 842 87
GenBank mRNA AF054817.1 - 4 - 5 832 87

View Full Product Details