Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DLEC1
Entrez Gene ID
9940
Gene Name
deleted in lung and esophageal cancer 1
Gene Aliases
CFAP81, DLC-1, DLC1, F56
Chromosome Location
Chr.3: 38038595 - 38122741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007337.3 NP_031363.2 35 - 36 5048 70
XM_006713438.3 XP_006713501.1 34 - 35 5504 70
XM_011534317.2 XP_011532619.1 35 - 36 5665 70
XM_011534318.2 XP_011532620.1 34 - 35 5492 70
GenBank mRNA AL137282.1 - 4 - 5 1111 70

View Full Product Details