The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133181.3 | NP_573444.2 | 14 - 15 | 1464 | 65 |
NM_139053.2 | NP_620641.1 | 14 - 15 | 1467 | 65 | |
XM_011542132.1 | XP_011540434.1 | 14 - 15 | 1389 | 65 | |
XM_011542133.1 | XP_011540435.1 | 14 - 15 | 1386 | 65 | |
XM_011542134.2 | XP_011540436.1 | 13 - 14 | 1388 | 65 | |
XM_017002327.1 | XP_016857816.1 | 14 - 15 | 1389 | 65 | |
GenBank mRNA | AK292198.1 | - | 14 - 15 | 1388 | 65 |
AY074930.1 | - | 14 - 15 | 1350 | 65 | |
BC012926.1 | - | 14 - 15 | 1387 | 65 | |
BM824107.1 | - | 3 - 4 | 157 | 65 |