The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173602.2 | NP_775873.2 | 34 - 35 | 4263 | 85 |
XM_005269044.1 | XP_005269101.1 | 34 - 35 | 4266 | 85 | |
XM_006719520.3 | XP_006719583.1 | 35 - 36 | 4239 | 85 | |
XM_006719521.3 | XP_006719584.1 | 34 - 35 | 4076 | 85 | |
XM_011538594.2 | XP_011536896.1 | 35 - 36 | 4292 | 85 | |
XM_011538595.2 | XP_011536897.1 | 33 - 34 | 4124 | 85 | |
XM_011538596.2 | XP_011536898.1 | 34 - 35 | 4079 | 85 | |
XM_017019699.1 | XP_016875188.1 | 35 - 36 | 4274 | 85 | |
XM_017019700.1 | XP_016875189.1 | 34 - 35 | 4248 | 85 | |
XM_017019701.1 | XP_016875190.1 | 34 - 35 | 4245 | 85 | |
XM_017019702.1 | XP_016875191.1 | 32 - 33 | 4095 | 85 | |
XM_017019703.1 | XP_016875192.1 | 33 - 34 | 4042 | 85 | |
XM_017019704.1 | XP_016875193.1 | 33 - 34 | 4050 | 85 | |
XM_017019705.1 | XP_016875194.1 | 33 - 34 | 4032 | 85 | |
GenBank mRNA | AB040896.2 | - | 25 - 26 | 2877 | 85 |
AK097369.1 | - | 16 - 17 | 1819 | 85 | |
AK126758.1 | - | 23 - 24 | 2634 | 85 | |
BC007671.2 | - | 18 - 19 | 2187 | 85 | |
BC030156.2 | - | 18 - 19 | 2187 | 85 |