Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
-
Chromosome Location
Chr.12: 50504985 - 50748667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173602.2 NP_775873.2 35 - 36 4326 101
XM_005269044.1 XP_005269101.1 35 - 36 4329 101
XM_006719520.3 XP_006719583.1 36 - 37 4302 101
XM_006719521.3 XP_006719584.1 35 - 36 4139 101
XM_011538594.2 XP_011536896.1 36 - 37 4355 101
XM_011538595.2 XP_011536897.1 34 - 35 4187 101
XM_011538596.2 XP_011536898.1 35 - 36 4142 101
XM_017019699.1 XP_016875188.1 36 - 37 4337 101
XM_017019700.1 XP_016875189.1 35 - 36 4311 101
XM_017019701.1 XP_016875190.1 35 - 36 4308 101
XM_017019702.1 XP_016875191.1 33 - 34 4158 101
XM_017019703.1 XP_016875192.1 34 - 35 4105 101
XM_017019704.1 XP_016875193.1 34 - 35 4113 101
XM_017019705.1 XP_016875194.1 34 - 35 4095 101
GenBank mRNA AB040896.2 - 26 - 27 2940 101
AK097369.1 - 17 - 18 1882 101
AK126758.1 - 24 - 25 2697 101
BC007671.2 - 19 - 20 2250 101
BC030156.2 - 19 - 20 2250 101

View Full Product Details