The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288834.1 | NP_001275763.1 | 2 - 3 | 912 | 67 |
NM_032207.3 | NP_115583.1 | 2 - 3 | 912 | 67 | |
XM_006722920.3 | XP_006722983.1 | 2 - 3 | 912 | 67 | |
XM_011528354.2 | XP_011526656.1 | 2 - 3 | 912 | 67 | |
XM_017027350.1 | XP_016882839.1 | 2 - 3 | 912 | 67 | |
GenBank mRNA | AK025395.1 | - | 2 - 3 | 810 | 67 |
AK225118.1 | - | 2 - 3 | 829 | 67 | |
AK225678.1 | - | 2 - 3 | 822 | 67 | |
BC027869.1 | - | 2 - 3 | 784 | 67 |