Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BST2
Entrez Gene ID
684
Gene Name
bone marrow stromal cell antigen 2
Gene Aliases
CD317, TETHERIN
Chromosome Location
Chr.19: 17402939 - 17405648 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004335.3 NP_004326.1 1 - 2 366 93
GenBank mRNA AK223124.1 - 1 - 2 304 93
AK291099.1 - 1 - 2 311 93
AK303593.1 - 1 - 2 336 93
BC033873.1 - 1 - 2 366 93
D28137.1 - 1 - 2 294 93

View Full Product Details