The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160227.1 | NP_001153699.1 | 2 - 3 | 465 | 154 |
NM_025137.3 | NP_079413.3 | 2 - 3 | 465 | 154 | |
XM_006720700.1 | XP_006720763.1 | 2 - 3 | 465 | 154 | |
XM_006720701.3 | XP_006720764.1 | 2 - 3 | 465 | 154 | |
XM_017022634.1 | XP_016878123.1 | 2 - 3 | 465 | 154 | |
XM_017022635.1 | XP_016878124.1 | 2 - 3 | 465 | 154 | |
GenBank mRNA | AB058743.2 | - | 2 - 3 | 435 | 154 |
AB470308.1 | - | 2 - 3 | 434 | 154 | |
AB470309.1 | - | 2 - 3 | 434 | 154 | |
AB470310.1 | - | 2 - 3 | 434 | 154 | |
AL834168.1 | - | 1 - 2 | 187 | 154 | |
BC047416.1 | - | 2 - 3 | 445 | 154 | |
BC150640.1 | - | 2 - 3 | 462 | 154 | |
BC153879.1 | - | 2 - 3 | 435 | 154 |