The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289178.1 | NP_001276107.1 | 1 - 2 | 227 | 64 |
GenBank mRNA | AJ577571.1 | - | 1 - 2 | 175 | 64 |
BC022027.1 | - | 1 - 2 | 203 | 64 |