Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDX18
Entrez Gene ID
8886
Gene Name
DEAD-box helicase 18
Gene Aliases
MrDb
Chromosome Location
Chr.2: 117814679 - 117832377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006773.3 NP_006764.3 3 - 4 613 128
GenBank mRNA AK001467.1 - 3 - 4 585 128
AK091227.1 - 1 - 2 78 128
BC001238.1 - 3 - 4 542 128
BC003360.1 - 3 - 4 572 128
BC024739.1 - 3 - 4 587 128
CR457060.1 - 2 - 3 334 128
X98743.1 - 2 - 3 351 128

View Full Product Details