Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRCA1
Entrez Gene ID
672
Gene Name
BRCA1, DNA repair associated
Gene Aliases
BRCAI, BRCC1, BROVCA1, FANCS, IRIS, PNCA4, PPP1R53, PSCP, RNF53
Chromosome Location
Chr.17: 43044295 - 43125483 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007299.3 NP_009230.2 1 - 2 176 134
GenBank mRNA BC072418.1 - 1 - 2 176 134
BC085615.1 - 1 - 2 84 134

View Full Product Details