The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004540.3 | NP_004531.2 | 13 - 14 | 2021 | 94 |
XM_011529575.2 | XP_011527877.1 | 14 - 15 | 2496 | 94 | |
XM_011529576.2 | XP_011527878.1 | 14 - 15 | 2073 | 94 | |
XM_011529579.2 | XP_011527881.1 | 13 - 14 | 1997 | 94 | |
XM_011529580.2 | XP_011527882.1 | 14 - 15 | 2496 | 94 | |
XM_011529581.2 | XP_011527883.1 | 14 - 15 | 2496 | 94 | |
XM_011529582.2 | XP_011527884.1 | 14 - 15 | 2495 | 94 | |
XM_011529585.2 | XP_011527887.1 | 10 - 11 | 1363 | 94 | |
XM_017028353.1 | XP_016883842.1 | 14 - 15 | 2161 | 94 | |
XM_017028354.1 | XP_016883843.1 | 13 - 14 | 1997 | 94 | |
XM_017028355.1 | XP_016883844.1 | 13 - 14 | 1997 | 94 | |
XM_017028356.1 | XP_016883845.1 | 13 - 14 | 1896 | 94 | |
XM_017028357.1 | XP_016883846.1 | 13 - 14 | 1896 | 94 | |
GenBank mRNA | AK299054.1 | - | 10 - 11 | 1534 | 94 |
BC052946.1 | - | 13 - 14 | 2021 | 94 | |
U75330.1 | - | 13 - 14 | 1841 | 94 |