The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004540.3 | NP_004531.2 | 5 - 6 | 871 | 94 |
XM_011529575.2 | XP_011527877.1 | 6 - 7 | 1346 | 94 | |
XM_011529576.2 | XP_011527878.1 | 6 - 7 | 923 | 94 | |
XM_011529579.2 | XP_011527881.1 | 5 - 6 | 847 | 94 | |
XM_011529580.2 | XP_011527882.1 | 6 - 7 | 1346 | 94 | |
XM_011529581.2 | XP_011527883.1 | 6 - 7 | 1346 | 94 | |
XM_011529582.2 | XP_011527884.1 | 6 - 7 | 1345 | 94 | |
XM_011529585.2 | XP_011527887.1 | 2 - 3 | 213 | 94 | |
XM_017028353.1 | XP_016883842.1 | 6 - 7 | 1011 | 94 | |
XM_017028354.1 | XP_016883843.1 | 5 - 6 | 847 | 94 | |
XM_017028355.1 | XP_016883844.1 | 5 - 6 | 847 | 94 | |
XM_017028356.1 | XP_016883845.1 | 5 - 6 | 746 | 94 | |
XM_017028357.1 | XP_016883846.1 | 5 - 6 | 746 | 94 | |
XM_017028358.1 | XP_016883847.1 | 6 - 7 | 1350 | 94 | |
GenBank mRNA | AK299054.1 | - | 2 - 3 | 384 | 94 |
AK302870.1 | - | 6 - 7 | 850 | 94 | |
BC052946.1 | - | 5 - 6 | 871 | 94 | |
U75330.1 | - | 5 - 6 | 691 | 94 |