The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006251.5 | NP_006242.5 | 7 - 8 | 1313 | 148 |
NM_206907.3 | NP_996790.3 | 8 - 9 | 1358 | 148 | |
XM_006714482.1 | XP_006714545.1 | 6 - 7 | 1207 | 148 | |
XM_017009624.1 | XP_016865113.1 | 8 - 9 | 2269 | 148 | |
XM_017009625.1 | XP_016865114.1 | 7 - 8 | 2214 | 148 | |
GenBank mRNA | AB022017.1 | - | 7 - 8 | 1303 | 148 |
AF100763.1 | - | 7 - 8 | 1283 | 148 | |
AK307546.1 | - | 8 - 9 | 1340 | 148 | |
AK312947.1 | - | 7 - 8 | 1295 | 148 | |
BC037303.2 | - | 7 - 8 | 1295 | 148 | |
BC048980.1 | - | 8 - 9 | 1537 | 148 | |
Y12856.1 | - | 1 - 2 | 401 | 148 |