The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282873.1 | NP_001269802.1 | 11 - 12 | 1783 | 142 |
NM_015902.5 | NP_056986.2 | 11 - 12 | 1783 | 142 | |
XM_011517104.2 | XP_011515406.2 | 10 - 11 | 1768 | 142 | |
XM_011517105.2 | XP_011515407.2 | 10 - 11 | 1768 | 142 | |
XM_011517106.2 | XP_011515408.2 | 10 - 11 | 1750 | 142 | |
XM_017013533.1 | XP_016869022.1 | 10 - 11 | 1768 | 142 | |
XM_017013534.1 | XP_016869023.1 | 10 - 11 | 1768 | 142 | |
GenBank mRNA | AB020703.3 | - | 11 - 12 | 1783 | 142 |
AF006010.2 | - | 11 - 12 | 1361 | 142 | |
BC137234.1 | - | 11 - 12 | 1355 | 142 | |
U95000.1 | - | 11 - 12 | 1325 | 142 |