The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282873.1 | NP_001269802.1 | 27 - 28 | 4027 | 116 |
NM_015902.5 | NP_056986.2 | 27 - 28 | 4027 | 116 | |
XM_005250962.4 | XP_005251019.1 | 6 - 7 | 788 | 116 | |
XM_011517104.2 | XP_011515406.2 | 26 - 27 | 4012 | 116 | |
XM_011517105.2 | XP_011515407.2 | 26 - 27 | 4012 | 116 | |
XM_011517106.2 | XP_011515408.2 | 26 - 27 | 3994 | 116 | |
XM_017013533.1 | XP_016869022.1 | 26 - 27 | 4012 | 116 | |
XM_017013534.1 | XP_016869023.1 | 26 - 27 | 4012 | 116 | |
GenBank mRNA | AB020703.3 | - | 27 - 28 | 4027 | 116 |
AF006010.2 | - | 27 - 28 | 3605 | 116 | |
BC137234.1 | - | 27 - 28 | 3599 | 116 | |
U95000.1 | - | 27 - 28 | 3569 | 116 |