Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLCA2
Entrez Gene ID
120376
Gene Name
colorectal cancer associated 2
Gene Aliases
C11orf93, CASC13, LOH11CR1G
Chromosome Location
Chr.11: 111298517 - 111308735 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136105.2 NP_001129577.1 3 - 4 621 65
NM_001271457.1 NP_001258386.1 3 - 4 366 65
NM_001271458.1 NP_001258387.1 3 - 4 190 65
XM_017017196.1 XP_016872685.1 4 - 5 270 65
XM_017017197.1 XP_016872686.1 2 - 3 1361 65
XM_017017198.1 XP_016872687.1 3 - 4 226 65
XM_017017199.1 XP_016872688.1 3 - 4 389 65
XM_017017200.1 XP_016872689.1 3 - 4 674 65
GenBank mRNA AK292195.1 - 3 - 4 621 65
BC042557.1 - 3 - 4 432 65
BC110079.1 - 2 - 3 89 65
BQ893717.1 - - 396 65
HY114242.1 - 3 - 4 191 65
KF740510.1 - 3 - 4 566 65
KF740511.1 - 3 - 4 181 65
KF740512.1 - 4 - 5 286 65
KF740513.1 - 4 - 5 412 65
KF740514.1 - 4 - 5 764 65

View Full Product Details