The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003061.2 | NP_003052.2 | 15 - 16 | 1744 | 63 |
GenBank mRNA | AB011537.3 | - | 15 - 16 | 1744 | 63 |
AB017167.1 | - | 15 - 16 | 1724 | 63 | |
AY029183.1 | - | 15 - 16 | 1471 | 63 | |
BC146761.1 | - | 15 - 16 | 1744 | 63 | |
BC146851.1 | - | 15 - 16 | 1657 | 63 |