The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003061.2 | NP_003052.2 | 31 - 32 | 3590 | 122 |
GenBank mRNA | AB011537.3 | - | 31 - 32 | 3590 | 122 |
AB017167.1 | - | 31 - 32 | 3570 | 122 | |
BC146761.1 | - | 31 - 32 | 3590 | 122 | |
BC146851.1 | - | 31 - 32 | 3503 | 122 |