The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003061.2 | NP_003052.2 | 35 - 36 | 4401 | 103 |
GenBank mRNA | AB011537.3 | - | 35 - 36 | 4401 | 103 |
AB017167.1 | - | 35 - 36 | 4381 | 103 | |
BC146761.1 | - | 35 - 36 | 4401 | 103 | |
BC146851.1 | - | 35 - 36 | 4314 | 103 |