The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271769.1 | NP_001258698.1 | 13 - 14 | 1548 | 96 |
NM_003664.4 | NP_003655.3 | 13 - 14 | 1536 | 96 | |
XM_005248618.3 | XP_005248675.1 | 13 - 14 | 1508 | 96 | |
XM_005248619.4 | XP_005248676.1 | 13 - 14 | 1508 | 96 | |
XM_017010001.1 | XP_016865490.1 | 13 - 14 | 1288 | 96 | |
GenBank mRNA | AK225908.1 | - | 13 - 14 | 1412 | 96 |
AK291201.1 | - | 13 - 14 | 1477 | 96 | |
AK303300.1 | - | 11 - 12 | 1156 | 96 | |
AY623424.1 | - | 13 - 14 | 1438 | 96 | |
BC038444.1 | - | 13 - 14 | 1497 | 96 | |
BX538041.1 | - | 13 - 14 | 1486 | 96 | |
KU178426.1 | - | 13 - 14 | 1360 | 96 | |
KU178427.1 | - | 13 - 14 | 1360 | 96 | |
U81504.1 | - | 13 - 14 | 1452 | 96 | |
U91931.1 | - | 13 - 14 | 1413 | 96 |