The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271769.1 | NP_001258698.1 | 20 - 21 | 2583 | 150 |
NM_003664.4 | NP_003655.3 | 20 - 21 | 2571 | 150 | |
XM_005248618.3 | XP_005248675.1 | 20 - 21 | 2543 | 150 | |
XM_005248619.4 | XP_005248676.1 | 20 - 21 | 2543 | 150 | |
XM_017010001.1 | XP_016865490.1 | 20 - 21 | 2323 | 150 | |
GenBank mRNA | AK225908.1 | - | 20 - 21 | 2447 | 150 |
AK291201.1 | - | 20 - 21 | 2512 | 150 | |
BC038444.1 | - | 20 - 21 | 2532 | 150 | |
BX538041.1 | - | 20 - 21 | 2521 | 150 | |
KU178426.1 | - | 20 - 21 | 2395 | 150 | |
U81504.1 | - | 20 - 21 | 2487 | 150 | |
U91931.1 | - | 20 - 21 | 2448 | 147 |