The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001156474.1 | NP_001149946.1 | 11 - 12 | 1819 | 146 |
NM_021827.4 | NP_068599.3 | 10 - 11 | 1549 | 146 | |
XM_005274161.1 | XP_005274218.1 | 10 - 11 | 1675 | 146 | |
XM_005274162.3 | XP_005274219.1 | 6 - 7 | 720 | 146 | |
XM_017018105.1 | XP_016873594.1 | 9 - 10 | 1173 | 146 | |
GenBank mRNA | AK027167.1 | - | 6 - 7 | 847 | 146 |
AK131331.1 | - | 10 - 11 | 1552 | 146 | |
AK223168.1 | - | 6 - 7 | 853 | 146 | |
AK298875.1 | - | 7 - 8 | 868 | 146 | |
BC126412.1 | - | 11 - 12 | 1663 | 146 |