The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017966.4 | NP_060436.4 | 1 - 2 | 53 | 116 |
GenBank mRNA | AK302384.1 | - | 1 - 2 | 121 | 116 |
AL834261.1 | - | 1 - 2 | 61 | 116 |