Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO45
Entrez Gene ID
200933
Gene Name
F-box protein 45
Gene Aliases
Fbx45
Chromosome Location
Chr.3: 196568854 - 196589059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105573.1 NP_001099043.1 1 - 2 453 63
GenBank mRNA BE778263.1 - 1 - 2 102 63
DB636510.1 - 1 - 2 456 63

View Full Product Details