Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX11A
Entrez Gene ID
8800
Gene Name
peroxisomal biogenesis factor 11 alpha
Gene Aliases
PEX11-ALPHA, PMP28, hsPEX11p
Chromosome Location
Chr.15: 89681531 - 89690784 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003847.2 NP_003838.1 2 - 3 328 100
GenBank mRNA AB015594.1 - 2 - 3 271 100
AF093668.1 - 2 - 3 233 100
AK001415.1 - 2 - 3 321 100
AK313318.1 - 2 - 3 242 100
BC009697.1 - 2 - 3 222 100
BC101164.2 - 2 - 3 182 100
BC114384.1 - 2 - 3 182 100
CR542046.1 - 2 - 3 176 100
CR542075.1 - 2 - 3 176 100

View Full Product Details