The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040194.1 | NP_001035284.1 | 1 - 2 | 153 | 58 |
NM_001040196.1 | NP_001035286.1 | 1 - 2 | 153 | 58 | |
NM_001040197.1 | NP_001035287.1 | 1 - 2 | 153 | 58 | |
NM_020350.4 | NP_065083.3 | 1 - 2 | 153 | 58 | |
GenBank mRNA | AF165187.1 | - | 1 - 2 | 29 | 58 |
AF312374.1 | - | 1 - 2 | 84 | 58 | |
AK290042.1 | - | 1 - 2 | 96 | 58 | |
BC011946.1 | - | 1 - 2 | 59 | 58 | |
BC017328.2 | - | 1 - 2 | 91 | 58 | |
BE782705.1 | - | - | 106 | 58 | |
BI117521.1 | - | 1 - 2 | 125 | 58 | |
CD674964.1 | - | 1 - 2 | 33 | 58 |