Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERCC5
Entrez Gene ID
2073
Gene Name
ERCC excision repair 5, endonuclease
Gene Aliases
COFS3, ERCC5-201, ERCM2, UVDR, XPG, XPGC
Chromosome Location
Chr.13: 102845841 - 102876001 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK294708.1 - 1 - 2 448 66
AK303558.1 - 1 - 2 306 66

View Full Product Details