Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARD3B
Entrez Gene ID
117583
Gene Name
par-3 family cell polarity regulator beta
Gene Aliases
ALS2CR19, PAR3B, PAR3L, PAR3beta
Chromosome Location
Chr.2: 204545793 - 205620162 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302769.1 NP_001289698.1 6 - 7 886 78
NM_057177.6 NP_476518.4 6 - 7 886 78
NM_152526.5 NP_689739.4 6 - 7 886 78
NM_205863.3 NP_995585.2 6 - 7 886 78
XM_011510552.2 XP_011508854.1 9 - 10 1229 78
XM_011510553.2 XP_011508855.1 8 - 9 5952 78
XM_017003283.1 XP_016858772.1 5 - 6 1176 78
XM_017003284.1 XP_016858773.1 6 - 7 4222 78
XM_017003285.1 XP_016858774.1 8 - 9 5959 78
XM_017003286.1 XP_016858775.1 5 - 6 4118 78
XM_017003287.1 XP_016858776.1 8 - 9 5955 78
XM_017003288.1 XP_016858777.1 8 - 9 5957 78
XM_017003289.1 XP_016858778.1 3 - 4 333 78
XM_017003290.1 XP_016858779.1 7 - 8 833 78
XM_017003291.1 XP_016858780.1 6 - 7 705 78
XM_017003292.1 XP_016858781.1 6 - 7 767 78
XM_017003293.1 XP_016858782.1 3 - 4 271 78
XM_017003294.1 XP_016858783.1 8 - 9 5949 78
GenBank mRNA AB073472.1 - 8 - 9 679 78
AB092439.1 - 8 - 9 679 78
AF428250.1 - 6 - 7 679 78
AF428251.1 - 6 - 7 679 78
AF466152.1 - 6 - 7 679 78
AK056157.1 - 8 - 9 768 78
AK057965.1 - 1 - 2 44 78

View Full Product Details