Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PARD3B
- Entrez Gene ID
-
117583
- Gene Name
- par-3 family cell polarity regulator beta
- Gene Aliases
-
ALS2CR19, PAR3B, PAR3L, PAR3beta
- Chromosome Location
-
Chr.2: 204545793 - 205620162 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057177.6 |
NP_476518.4 |
16 - 17 |
2396 |
87 |
View Full Product Details