Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INO80D
Entrez Gene ID
54891
Gene Name
INO80 complex subunit D
Gene Aliases
-
Chromosome Location
Chr.2: 205993721 - 206086182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017759.4 NP_060229.3 4 - 5 1369 74
XM_011511369.2 XP_011509671.1 4 - 5 1362 74
XM_011511370.2 XP_011509672.1 4 - 5 1274 74
XM_011511371.2 XP_011509673.1 4 - 5 1201 74
XM_011511373.2 XP_011509675.1 4 - 5 1203 74
XM_011511375.2 XP_011509677.1 4 - 5 1087 74
XM_011511376.2 XP_011509678.1 4 - 5 1074 74
GenBank mRNA AK000316.1 - 1 - 2 690 74
AK096585.1 - 4 - 5 1370 74
AK225339.1 - 1 - 2 690 74
BC004193.2 - 3 - 4 1086 74
BC011687.2 - 3 - 4 1086 74
BC017290.2 - 3 - 4 1086 74
BC136278.1 - 3 - 4 1067 74

View Full Product Details