The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024312.4 | NP_077288.2 | 3 - 4 | 512 | 85 |
XM_006719593.2 | XP_006719656.1 | 3 - 4 | 507 | 85 | |
XM_011538731.2 | XP_011537033.1 | 3 - 4 | 508 | 85 | |
XM_017019961.1 | XP_016875450.1 | 3 - 4 | 409 | 85 | |
XM_017019962.1 | XP_016875451.1 | 3 - 4 | 491 | 85 | |
GenBank mRNA | AK056137.1 | - | 3 - 4 | 383 | 85 |
AM085438.1 | - | 3 - 4 | 499 | 85 | |
AY687932.1 | - | 3 - 4 | 484 | 85 | |
BC071687.1 | - | 3 - 4 | 460 | 85 | |
BC131787.1 | - | 3 - 4 | 440 | 85 | |
BP276948.1 | - | 3 - 4 | 512 | 85 |