Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIC8B
Entrez Gene ID
55188
Gene Name
RIC8 guanine nucleotide exchange factor B
Gene Aliases
RIC8, hSyn
Chromosome Location
Chr.12: 106774285 - 106889316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018157.2 NP_060627.2 1 - 2 212 141
XM_005268998.3 XP_005269055.1 1 - 2 238 141
XM_017019548.1 XP_016875037.1 1 - 2 238 141
GenBank mRNA AK001482.1 - 1 - 2 119 141
BC034220.1 - 1 - 2 162 234
BC132689.1 - 1 - 2 193 141
BC144056.1 - 1 - 2 193 141
DA325821.1 - 1 - 2 212 141

View Full Product Details