The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001093.3 | NP_001084.3 | 30 - 31 | 4352 | 64 |
XM_005253876.4 | XP_005253933.1 | 31 - 32 | 4462 | 64 | |
XM_006719367.3 | XP_006719430.1 | 30 - 31 | 3983 | 64 | |
XM_011538259.2 | XP_011536561.1 | 31 - 32 | 5100 | 64 | |
XM_011538263.2 | XP_011536565.1 | 30 - 31 | 4911 | 64 | |
XM_011538264.2 | XP_011536566.1 | 30 - 31 | 4437 | 64 | |
XM_017019252.1 | XP_016874741.1 | 29 - 30 | 3778 | 64 | |
GenBank mRNA | AB209110.1 | - | 20 - 21 | 2406 | 64 |
AJ575592.3 | - | 30 - 31 | 4343 | 64 | |
AK302469.1 | - | 3 - 4 | 281 | 64 | |
AK302502.1 | - | 4 - 5 | 414 | 64 | |
AY382667.1 | - | 31 - 32 | 4462 | 64 | |
BC028417.1 | - | 4 - 5 | 438 | 64 | |
DQ493870.1 | - | 30 - 31 | 4343 | 64 | |
U34591.1 | - | 4 - 5 | 374 | 64 |