The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001093.3 | NP_001084.3 | 3 - 4 | 932 | 122 |
XM_005253876.4 | XP_005253933.1 | 4 - 5 | 1042 | 122 | |
XM_006719367.3 | XP_006719430.1 | 3 - 4 | 563 | 122 | |
XM_011538259.2 | XP_011536561.1 | 4 - 5 | 1680 | 122 | |
XM_011538263.2 | XP_011536565.1 | 4 - 5 | 1680 | 122 | |
XM_011538264.2 | XP_011536566.1 | 3 - 4 | 1017 | 122 | |
XM_011538265.2 | XP_011536567.1 | 4 - 5 | 1682 | 122 | |
XM_017019252.1 | XP_016874741.1 | 3 - 4 | 547 | 122 | |
GenBank mRNA | AJ575431.2 | - | 3 - 4 | 923 | 122 |
AJ575592.3 | - | 3 - 4 | 923 | 122 | |
AY382667.1 | - | 4 - 5 | 1042 | 122 | |
DQ493870.1 | - | 3 - 4 | 923 | 122 |