The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152405.4 | NP_689618.4 | 9 - 10 | 3209 | 74 |
XM_005248430.2 | XP_005248487.1 | 9 - 10 | 3086 | 74 | |
XM_011543155.2 | XP_011541457.1 | 8 - 9 | 3113 | 74 | |
GenBank mRNA | AK095189.1 | - | 8 - 9 | 1610 | 74 |
BC130624.1 | - | 9 - 10 | 1599 | 74 | |
BX648731.1 | - | 1 - 2 | 407 | 74 |