The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152405.4 | NP_689618.4 | 7 - 8 | 2508 | 102 |
XM_005248430.2 | XP_005248487.1 | 7 - 8 | 2508 | 102 | |
GenBank mRNA | AK095189.1 | - | 6 - 7 | 909 | 102 |
BC130624.1 | - | 6 - 7 | 934 | 102 |