The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261400.2 | NP_001248329.1 | 3 - 4 | 454 | 63 |
NM_002813.6 | NP_002804.2 | 5 - 6 | 769 | 63 | |
NR_048555.2 | 4 - 5 | 557 | 63 | ||
GenBank mRNA | AK055339.1 | - | 1 - 2 | 1725 | 63 |
BC002383.2 | - | 5 - 6 | 729 | 63 | |
BC004184.2 | - | 5 - 6 | 729 | 63 | |
BC004213.2 | - | 5 - 6 | 729 | 63 | |
BM850893.1 | - | 4 - 5 | 478 | 63 | |
CB111716.1 | - | 3 - 4 | 399 | 63 |