The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003279.2 | NP_003270.1 | 2 - 3 | 120 | 79 |
XM_011529031.2 | XP_011527333.1 | 2 - 3 | 130 | 79 | |
GenBank mRNA | AK291323.1 | - | 2 - 3 | 118 | 79 |
AK300665.1 | - | - | 118 | 79 | |
BC005323.1 | - | 2 - 3 | 118 | 79 | |
BU854061.1 | - | 2 - 3 | 151 | 79 | |
CR541864.1 | - | 1 - 2 | 54 | 79 | |
CR541884.1 | - | 1 - 2 | 54 | 79 | |
M22307.1 | - | 2 - 3 | 101 | 79 | |
X07898.1 | - | 2 - 3 | 118 | 79 |